Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Itm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008409.2 | NP_032435.2 | 6 | 1461 | ||
GenBank mRNA | AK014024.1 | - | 6 | 1231 | ||
AK051956.1 | - | 6 | 1440 | |||
AK135229.1 | - | 6 | 1413 | |||
AK167351.1 | - | 6 | 1462 | |||
BC031420.1 | - | 6 | 1428 | 3668557 | ||
L38971.1 | - | 6 | 1464 |