Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008446.2 | NP_032472.2 | 28 | 3715 | ||
GenBank mRNA | AK084837.1 | - | 15 | 2051 | ||
AK135430.1 | - | 28 | 3210 | |||
AK136346.1 | - | 9 | 956 | |||
AK171938.1 | - | 28 | 3627 | |||
BC050946.1 | - | 28 | 3653 | 6822283 | ||
BC054537.1 | - | 28 | 3637 | |||
D12646.1 | - | 28 | 4042 |