Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008449.2 | NP_032475.2 | 26 | 5616 | ||
GenBank mRNA | AB093244.1 | - | 26 | 5508 | ||
BC067051.1 | - | 26 | 5616 | 30536079 |