Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008465.5 | NP_032491.2 | 14 | 1857 | ||
GenBank mRNA | AK028259.1 | - | 14 | 1858 | ||
AK028307.1 | - | 14 | 1876 | |||
AK141446.1 | - | 1 | 73 | |||
AK154800.1 | - | 14 | 1875 | |||
AK169311.1 | - | 14 | 1880 | |||
AK169348.1 | - | 14 | 1880 | |||
BC006771.1 | - | 14 | 1842 | 3588534 | ||
U20619.1 | - | 14 | 1862 |