Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltb4r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008519.2 | NP_032545.1 | 2 | 1363 | ||
GenBank mRNA | AF044030.1 | - | 2 | 1359 | ||
AF110104.1 | - | 2 | 1310 | |||
AK010895.1 | - | 2 | 1349 | |||
AK155181.1 | - | 3 | 1156 | |||
BC064063.1 | - | 2 | 1294 | 874601 |
Set Membership: |
![]() |