Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008528.4 | NP_032554.2 | 5 | 764 | ||
GenBank mRNA | AB015290.1 | - | 5 | 438 | ||
AF068182.1 | - | 5 | 419 | |||
AJ222814.1 | - | 5 | 389 | |||
AK089704.1 | - | 5 | 404 | |||
BC059785.1 | - | 5 | 390 | 5339723 | ||
Y17159.1 | - | 5 | 764 |