Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NUCB2 SIRNA Assays ›
Gene Symbol:
NUCB2
Gene Name:
nucleobindin 2
Gene Aliases:
HEL-S-109, NEFA
Chromosome Location:
Chr.11: 17260340 - 17331881 on Build GRCh38
UniGene ID:
Hs.654599
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15522
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NUCB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005013.2
NP_005004.1
4, 5
496
XM_017017804.1
XP_016873293.1
6, 7
731
XM_017017805.1
XP_016873294.1
5, 6
647
XM_017017806.1
XP_016873295.1
5, 6
590
XM_017017807.1
XP_016873296.1
5, 6
601
XM_017017808.1
XP_016873297.1
4, 5
535
XM_017017809.1
XP_016873298.1
4, 5
532
XM_017017810.1
XP_016873299.1
6, 7
1002
XM_017017811.1
XP_016873300.1
6, 7
911
XM_017017812.1
XP_016873301.1
4, 5
496
XM_017017813.1
XP_016873302.1
5, 6
595
XM_017017814.1
XP_016873303.1
5, 6
588
XM_017017815.1
XP_016873304.1
7, 8
995
XM_017017816.1
XP_016873305.1
5, 6
765
XM_017017817.1
XP_016873306.1
5, 6
690
XM_017017818.1
XP_016873307.1
3, 4
341
XM_017017819.1
XP_016873308.1
3, 4
286
XM_017017820.1
XP_016873309.1
6, 7
731
XM_017017821.1
XP_016873310.1
6, 7
727
XM_017017822.1
XP_016873311.1
4, 5
499
XM_017017823.1
XP_016873312.1
5, 6
603
XM_017017824.1
XP_016873313.1
5, 6
601
XM_017017825.1
XP_016873314.1
4, 5
519
XM_017017826.1
XP_016873315.1
4, 5
532
XM_017017827.1
XP_016873316.1
6, 7
958
XM_017017828.1
XP_016873317.1
5, 6
588
XM_017017829.1
XP_016873318.1
3, 4
364
GenBank mRNA
AB231311.1
-
2, 3
251
AF052642.1
-
2, 3
251
AF052643.1
-
2, 3
251
AF052644.1
-
2, 3
251
AF450266.1
-
4, 5
465
AK093209.1
-
10, 11
1492
AK097398.1
-
6, 7
912
AK128739.1
-
8, 9
2163
AK291507.1
-
4, 5
526
DA816251.1
-
4, 5
526
FJ224327.1
-
4, 5
470
X76732.1
-
4, 5
470
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Process(es)
DNA binding
calcium ion binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0