Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008539.3 | NP_032565.2 | 6 | 1532 | ||
GenBank mRNA | AK017583.1 | - | 6 | 1493 | ||
AK054104.1 | - | 6 | 1493 | |||
AK084505.1 | - | 6 | 1600 | |||
AK158509.1 | - | 6 | 1548 | |||
BC058693.1 | - | 6 | 1532 | 6811514 | ||
U58992.1 | - | 5 | 1205 | |||
U74359.1 | - | 6 | 1398 |