Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Smad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164041.1 | NP_001157513.1 | 2 | 997 | ||
NM_001164042.1 | NP_001157514.1 | 2 | 617 | |||
NM_008541.3 | NP_032567.1 | 2 | 594 | |||
GenBank mRNA | AF063006.1 | - | 2 | 998 | ||
AK018077.1 | - | 2 | 566 | |||
AK082997.1 | - | 2 | 618 | |||
AK139765.1 | - | 2 | 539 | |||
BC048233.1 | - | 2 | 505 | |||
BC050001.1 | - | 2 | 497 | 6490853 | ||
U58993.1 | - | 2 | 573 | |||
U77638.1 | - | 2 | 743 |