Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008563.2 | NP_032589.1 | 17 | 2848 | ||
GenBank mRNA | AK088142.1 | - | 16 | 3353 | ||
AK133850.1 | - | 17 | 2913 | |||
AK145563.1 | - | 17 | 2947 | |||
AK146497.1 | - | 17 | 2861 | |||
AK147065.1 | - | 17 | 2863 | |||
AK148006.1 | - | 17 | 2893 | |||
BC020400.1 | - | 17 | 2862 | |||
BC031700.1 | - | 17 | 2847 | 5007886 | ||
X62154.1 | - | 17 | 2728 |