Search Thermo Fisher Scientific
Gene Symbol: Mep1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008585.2 | NP_032611.2 | 8 | 726 | ||
GenBank mRNA | AK143906.1 | - | 8 | 684 | ||
AK164734.1 | - | 8 | 681 | |||
BC015258.1 | - | 8 | 726 | 4241252 | ||
M74897.1 | - | 8 | 725 |