Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mep1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008586.2 | NP_032612.2 | 10 | 1142 | ||
GenBank mRNA | AK098127.1 | - | 10 | 1142 | ||
BC125627.1 | - | 10 | 1140 | 40130142 | ||
BC145979.1 | - | 10 | 1141 | 40131282 | ||
L15193.1 | - | 11 | 1142 |