Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008609.3 | NP_032635.1 | 10 | 3358 | ||
GenBank mRNA | AK004523.1 | - | 1 | 262 | ||
BC047278.1 | - | 10 | 3358 | 6390113 | ||
BC057952.1 | - | 10 | 2928 | 5342023 | ||
D86332.1 | - | 11 | 2824 |