Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mmp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008611.4 | NP_032637.3 | 10 | 2041 | ||
GenBank mRNA | AK089234.1 | - | 10 | 2010 | ||
AK089336.1 | - | 10 | 2015 | |||
AK089381.1 | - | 10 | 2014 | |||
AK089516.1 | - | 10 | 2002 | |||
AK137468.1 | - | 10 | 2014 | |||
AK154937.1 | - | 10 | 2013 | |||
AK155832.1 | - | 10 | 2012 | |||
AK155901.1 | - | 10 | 2017 | |||
AK170177.1 | - | 10 | 2012 | |||
AK171168.1 | - | 10 | 2011 | |||
AK171613.1 | - | 10 | 2014 | |||
BY548872.1 | - | 1 | 44 |