Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APBA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004886.3 | NP_004877.1 | 2 | 614 | ||
XM_006722950.3 | XP_006723013.1 | 2 | 636 | |||
XM_006722951.3 | XP_006723014.1 | 1 | 361 | |||
GenBank mRNA | AB021638.1 | - | 2 | 604 | ||
BC086306.1 | - | 1 | 473 | 6371716 | ||
BI910038.1 | - | 2 | 616 |