Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008636.4 | NP_032662.3 | 11 | 2531 | ||
GenBank mRNA | AK030284.1 | - | 11 | 2510 | ||
AK036233.1 | - | 1 | 236 | |||
AK150295.1 | - | 12 | 2585 | |||
BC017679.1 | - | 11 | 2491 | 4210829 | ||
X71327.1 | - | 11 | 2475 |