Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008637.1 | NP_032663.1 | 5 | 711 | ||
GenBank mRNA | AK011695.1 | - | 5 | 700 | ||
AK088309.1 | - | 5 | 703 | |||
BC021940.1 | - | 4 | 682 | 5067377 | ||
BC098239.1 | - | 4 | 681 | 3469785 | ||
D49956.1 | - | 5 | 711 |