Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gadd45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008655.1 | NP_032681.1 | 4 | 719 | ||
GenBank mRNA | AK010420.1 | - | 4 | 737 | ||
AK149895.1 | - | 4 | 737 | |||
AK154952.1 | - | 4 | 737 | |||
AK186667.1 | - | 2 | 181 | |||
BC023815.1 | - | 4 | 731 | 5355150 | ||
X54149.1 | - | 4 | 719 |