Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008675.2 | NP_032701.2 | 4 | 1704 | ||
GenBank mRNA | AK133787.1 | - | 4 | 1703 | ||
AK146535.1 | - | 4 | 1718 | |||
AK158008.1 | - | 4 | 1713 | |||
AK167311.1 | - | 4 | 1702 | |||
D50263.1 | - | 4 | 1602 |