Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008682.2 | NP_032708.2 | 3 | 313 | ||
GenBank mRNA | AK010591.1 | - | 3 | 301 | ||
AK048393.1 | - | 3 | 265 | |||
AK085355.1 | - | 3 | 276 | |||
AK090040.1 | - | 3 | 258 | |||
AK153746.1 | - | 3 | 315 | |||
BC052430.1 | - | 3 | 282 | 6836583 | ||
BC066870.1 | - | 3 | 250 | 30476383 | ||
D10712.1 | - | 3 | 255 |