Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008691.2 | NP_032717.2 | 3 | 1694 | ||
GenBank mRNA | AK018234.1 | - | 3 | 1533 | ||
AK051296.1 | - | 3 | 1537 | |||
AK080794.1 | - | 3 | 1538 | |||
AK164318.1 | - | 3 | 1537 | |||
AK165041.1 | - | 3 | 1537 | |||
BC052352.1 | - | 3 | 1536 | |||
BC058333.1 | - | 3 | 1527 | |||
BC119602.2 | - | 3 | 1502 | |||
BC128564.1 | - | 3 | 1497 | |||
DQ201636.1 | - | 3 | 1512 | |||
M20481.1 | - | 3 | 540 |