Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008703.2 | NP_032729.1 | 2 | 564 | ||
GenBank mRNA | BC119237.1 | - | 2 | 633 | 8733986 | |
BC144884.1 | - | 2 | 634 |
Set Membership: |
![]() |