Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290368.1 | NP_001277297.1 | 11 | 1213 | ||
NM_001290369.1 | NP_001277298.1 | 10 | 1195 | |||
NM_001290370.1 | NP_001277299.1 | 9 | 1152 | |||
NM_008708.2 | NP_032734.1 | 11 | 1245 | |||
GenBank mRNA | AF043327.1 | - | 11 | 1169 | ||
AK134521.1 | - | 11 | 1209 | |||
AK161490.1 | - | 11 | 1213 | |||
AK168171.1 | - | 11 | 1213 | |||
AK168190.1 | - | 11 | 1213 | |||
BC037647.1 | - | 10 | 1142 | 5253732 |