Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008711.2 | NP_032737.1 | 1 | 700 | ||
GenBank mRNA | AK146488.1 | - | 1 | 703 | ||
BC132527.1 | - | 1 | 226 | 40130804 | ||
BC138148.1 | - | 1 | 226 | 8861167 |