Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136074.2 | NP_001129546.1 | 4 | 1114 | ||
NM_001271916.1 | NP_001258845.1 | 4 | 1134 | |||
NM_001271917.1 | NP_001258846.1 | 3 | 1007 | |||
NM_008736.3 | NP_032762.1 | 3 | 1113 | |||
GenBank mRNA | AK044244.1 | - | 4 | 1135 | ||
AK044275.1 | - | 3 | 1078 | |||
AK044316.1 | - | 4 | 1097 | |||
BC031440.1 | - | 4 | 1098 | 4527590 | ||
BC051241.1 | - | 1 | 4145 | |||
L14935.1 | - | 3 | 1052 |