Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195046.1 | NP_001181975.1 | 11 | 1055 | ||
NM_001195047.1 | NP_001181976.1 | 10 | 1010 | |||
NM_001195048.1 | NP_001181977.1 | 11 | 1165 | |||
NM_001195049.1 | NP_001181978.1 | 13 | 1429 | |||
NM_008778.3 | NP_032804.2 | 10 | 1038 | |||
GenBank mRNA | AF082297.1 | - | 9 | 952 | ||
AJ496262.1 | - | 9 | 920 | |||
AJ496263.1 | - | 10 | 965 | |||
AK158893.1 | - | 11 | 1165 | |||
AM943853.1 | - | 10 | 983 | |||
AM943854.1 | - | 11 | 1028 | |||
BC053403.1 | - | 10 | 1045 | 30060082 | ||
U39738.1 | - | 14 | 1436 |