Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162946.1 | NP_001156418.1 | 4 | 402 | ||
NM_008797.3 | NP_032823.2 | 3 | 379 | |||
GenBank mRNA | AK128927.1 | - | 329 | |||
AK148328.1 | - | 4 | 349 | |||
AK170596.1 | - | 4 | 362 | |||
AK170871.1 | - | 4 | 352 | |||
AK172320.1 | - | 4 | 330 | |||
BC055030.1 | - | 4 | 337 | 5053475 | ||
BG087228.2 | - | 4 | 338 | |||
BY059816.1 | - | 3 | 380 | |||
BY139237.1 | - | 3 | 377 | |||
L09192.1 | - | 3 | 355 |