Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162946.1 | NP_001156418.1 | 4, 5 | 476 | ||
NM_008797.3 | NP_032823.2 | 3, 4 | 453 | |||
GenBank mRNA | AK128927.1 | - | 403 | |||
AK148328.1 | - | 4, 5 | 423 | |||
AK170596.1 | - | 4, 5 | 436 | |||
AK170871.1 | - | 4, 5 | 426 | |||
AK172320.1 | - | 4, 5 | 404 | |||
BC055030.1 | - | 4, 5 | 411 | 5053475 | ||
BG087228.2 | - | 4, 5 | 412 | |||
L09192.1 | - | 3, 4 | 429 |