Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162415.1 | NP_001155887.1 | 4, 5 | 447 | ||
NM_001162416.1 | NP_001155888.1 | 2, 3 | 240 | |||
NM_008825.4 | NP_032851.2 | 3, 4 | 341 | |||
GenBank mRNA | AK029659.1 | - | 4, 5 | 448 | ||
AK146744.1 | - | 3, 4 | 335 | |||
BC018418.1 | - | 3, 4 | 298 | 3979400 | ||
BC051014.1 | - | 2, 3 | 254 | 6493944 | ||
BY037226.1 | - | 3, 4 | 337 | |||
EU616666.1 | - | 2, 3 | 239 | |||
EU616667.1 | - | 2, 3 | 239 | |||
EU616668.1 | - | 2, 3 | 239 | |||
EU616669.1 | - | 2, 3 | 239 | |||
EU616670.1 | - | 2, 3 | 249 | |||
X98847.1 | - | 2, 3 | 259 |