Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162416.1 | NP_001155888.1 | 13 | 2098 | ||
NM_008825.4 | NP_032851.2 | 15 | 2262 | |||
GenBank mRNA | AK029556.1 | - | 1 | 108 | ||
AK146744.1 | - | 15 | 2256 | |||
BC018418.1 | - | 15 | 2219 | 3979400 | ||
BC051014.1 | - | 14 | 2175 | 6493944 |