Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 3, 4 | 520 | ||
NM_010589.6 | NP_034719.2 | 3, 4 | 520 | |||
GenBank mRNA | AK038268.1 | - | 2, 3 | 331 | ||
AK088365.1 | - | 3, 4 | 372 | |||
AK142178.1 | - | 3, 4 | 358 | |||
AK156646.1 | - | 3, 4 | 342 | |||
AK164520.1 | - | 3, 4 | 360 | |||
BC027234.1 | - | 351 | ||||
BC105577.1 | - | 2, 3 | 323 | 3968386 | ||
BC131646.1 | - | 2, 3 | 331 | 40110146 | ||
BC131647.1 | - | 2, 3 | 331 | 40110148 | ||
L32955.1 | - | 2, 3 | 322 | |||
L33768.1 | - | 2, 3 | 316 | |||
L40172.1 | - | 3, 4 | 381 |