Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111096.1 | NP_001104566.1 | 5 | 617 | ||
NM_010747.2 | NP_034877.2 | 5 | 554 | |||
GenBank mRNA | AK028112.1 | - | 5 | 550 | ||
AK151914.1 | - | 5 | 615 | |||
AK152199.1 | - | 5 | 615 | |||
AK153038.1 | - | 5 | 551 | |||
AK170561.1 | - | 5 | 615 | |||
BC031547.1 | - | 5 | 587 | 3589207 | ||
BC157998.1 | - | 5 | 386 | 9007404 | ||
BY743506.1 | - | 5 | 622 | |||
M57696.1 | - | 5 | 453 | |||
M57697.1 | - | 5 | 390 | |||
M64608.1 | - | 5 | 603 |