Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010764.2 | NP_034894.2 | 2, 3 | 315 | ||
GenBank mRNA | AK004817.2 | - | 2, 3 | 272 | ||
AK035551.1 | - | 2, 3 | 270 | |||
AK088826.1 | - | 2, 3 | 273 | |||
AK147928.1 | - | 2, 3 | 273 | |||
AK171166.1 | - | 2, 3 | 273 | |||
BC005430.1 | - | 2, 3 | 260 | 3488681 | ||
U29947.1 | - | 2, 3 | 258 | |||
U87240.1 | - | 2, 3 | 249 |