Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ITM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171581.1 | NP_001165052.1 | 4 | 822 | ||
NM_004867.4 | NP_004858.1 | 5 | 954 | |||
GenBank mRNA | AF038953.1 | - | 5 | 756 | ||
AK223327.1 | - | 5 | 733 | |||
AK303238.1 | - | 4 | 604 | |||
AK313154.1 | - | 5 | 912 | |||
AY359051.1 | - | 5 | 695 | |||
BC040437.1 | - | 5 | 709 | 5748519 | ||
BM561934.1 | - | 2 | 197 | |||
CR456875.1 | - | 5 | 617 |