Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ncoa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010881.2 | NP_035011.1 | 23 | 5120 | ||
GenBank mRNA | AK086638.1 | - | 1 | 40 | ||
AK154861.1 | - | 1 | 178 | |||
BC068177.1 | - | 20 | 4428 | 30628581 | ||
BC080866.1 | - | 23 | 4797 | 30532122 | ||
U56920.1 | - | 22 | 4678 | |||
U64606.1 | - | 19 | 4368 | |||
U64828.1 | - | 23 | 5033 |