Gene Symbol: Nfe2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010902.3 | NP_035032.1 | 5 | 1020 | ||
GenBank mRNA | AK142347.1 | - | 5 | 922 | ||
AK144247.1 | - | 5 | 894 | |||
BC003340.1 | - | 5 | 876 | |||
BC026943.1 | - | 5 | 961 | 3663276 | ||
U20532.1 | - | 5 | 1036 |