Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122952.1 | NP_001116424.1 | 2 | 445 | ||
NM_001122953.1 | NP_001116425.1 | 2 | 684 | |||
NM_010905.3 | NP_035035.1 | 2 | 684 | |||
GenBank mRNA | AF326553.1 | - | 2 | 383 | ||
AF326554.1 | - | 2 | 383 | |||
AK052204.1 | - | 2 | 506 | |||
AK054310.1 | - | 2 | 548 | |||
AK137731.1 | - | 2 | 547 | |||
AK139357.1 | - | 2 | 446 | |||
AK173173.1 | - | 1 | 320 | |||
BC075702.1 | - | 2 | 665 | 30087137 | ||
BC122878.1 | - | 2 | 406 | 40107695 | ||
BC122880.1 | - | 2 | 404 | |||
D90172.1 | - | 2 | 614 | |||
D90173.1 | - | 2 | 522 | |||
D90174.1 | - | 2 | 555 | |||
D90175.1 | - | 399 | ||||
D90176.1 | - | 2 | 437 | |||
U57633.1 | - | 2 | 339 | |||
Y07690.1 | - | 2 | 415 | |||
Y07691.1 | - | 2 | 413 |