Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010907.2 | NP_035037.2 | 2 | 398 | ||
GenBank mRNA | AK089215.1 | - | 2 | 347 | ||
AK151115.1 | - | 2 | 351 | |||
AK151608.1 | - | 2 | 351 | |||
AK161622.1 | - | 2 | 310 | |||
BC046754.1 | - | 2 | 331 | 5693175 | ||
U36277.1 | - | 2 | 342 |