Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252282.1 | NP_001239211.1 | 2 | 213 | ||
NM_001252283.1 | NP_001239212.1 | 2 | 213 | |||
NM_001252287.1 | NP_001239216.1 | 2 | 289 | |||
NM_001252288.1 | NP_001239217.1 | 2 | 273 | |||
NM_010956.4 | NP_035086.2 | 2 | 213 | |||
GenBank mRNA | AK008811.1 | - | 2 | 169 | ||
AK147289.1 | - | 2 | 214 | |||
AK150009.1 | - | 2 | 179 | |||
AK169286.1 | - | 2 | 205 | |||
AK220536.1 | - | 2 | 175 | |||
BC013670.1 | - | 2 | 147 | 4188988 | ||
BC049104.1 | - | 2 | 226 | 6417392 | ||
BC057354.1 | - | 2 | 141 | 6827509 | ||
BC061232.1 | - | 2 | 169 | |||
BY768178.1 | - | 2 | 206 | |||
BY785860.1 | - | 2 | 270 |