Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011020.3 | NP_035150.3 | 7 | 787 | ||
GenBank mRNA | AB001926.1 | - | 7 | 753 | ||
AK144225.1 | - | 7 | 824 | |||
AK163459.1 | - | 8 | 959 | |||
BC012712.1 | - | 7 | 856 | 4190161 | ||
BC057002.1 | - | 7 | 787 | 5702602 | ||
BC110662.1 | - | 7 | 807 | 4511154 | ||
D49482.1 | - | 7 | 781 | |||
U23921.1 | - | 7 | 904 |