Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004984.2 | NP_004975.2 | 12, 13 | 1509 | ||
GenBank mRNA | AB210045.1 | - | 12, 13 | 1334 | ||
AB290174.1 | - | 12, 13 | 1575 | |||
AK294881.1 | - | 9, 10 | 1262 | |||
AK315202.1 | - | 12, 13 | 1529 | |||
BC146670.1 | - | 12, 13 | 1512 | 40148192 | ||
BC150208.1 | - | 12, 13 | 1512 | 40148890 | ||
U06698.1 | - | 12, 13 | 1449 |