Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113213.1 | NP_001106684.1 | 13 | 1641 | ||
NM_001113214.1 | NP_001106685.1 | 13 | 1641 | |||
NM_001177659.1 | NP_001171130.1 | 13 | 1644 | |||
NM_001177661.1 | NP_001171132.1 | 12 | 1487 | |||
NM_013615.3 | NP_038643.1 | 13 | 1614 | |||
GenBank mRNA | AF000968.1 | - | 11 | 1461 | ||
AF034105.1 | - | 13 | 1577 | |||
AK143726.1 | - | 13 | 1548 | |||
AK148523.1 | - | 13 | 1554 | |||
AK168415.1 | - | 13 | 1543 | |||
BC057001.1 | - | 14 | 1698 | 6827260 | ||
BC067406.1 | - | 3 | 331 |