Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013626.3 | NP_038654.2 | 3 | 607 | ||
GenBank mRNA | AK049423.1 | - | 2 | 268 | ||
AK051649.1 | - | 3 | 597 | |||
U79523.1 | - | 3 | 492 |