Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013626.3 | NP_038654.2 | 14, 15 | 1631 | ||
GenBank mRNA | U79523.1 | - | 14, 15 | 1516 |