Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013822.5 | NP_038850.1 | 26 | 5152 | ||
GenBank mRNA | AK078018.1 | - | 3 | 1919 | ||
AK137092.1 | - | 26 | 5049 | |||
BC058675.1 | - | 26 | 4966 | 6834418 |