Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109040.2 | NP_001102510.1 | 3 | 651 | ||
NM_001109041.2 | NP_001102511.1 | 3 | 651 | |||
NM_001109042.2 | NP_001102512.1 | 3 | 651 | |||
NM_016705.4 | NP_057914.2 | 3 | 651 | |||
GenBank mRNA | AF202892.1 | - | 3 | 511 | ||
AK008213.1 | - | 3 | 540 | |||
AK019525.1 | - | 3 | 652 | |||
AK047350.1 | - | 3 | 541 | |||
AK047880.1 | - | 3 | 540 | |||
AK147436.1 | - | 3 | 539 | |||
AK163536.1 | - | 3 | 535 | |||
BC060698.1 | - | 3 | 518 | 6409819 | ||
BC062896.1 | - | 3 | 518 | 6409819 | ||
BY714299.1 | - | 3 | 648 |