Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy5r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016708.3 | NP_057917.2 | 2 | 317 | ||
GenBank mRNA | AB001346.1 | - | 2 | 320 | ||
AF049329.1 | - | 1 | 188 | |||
AK045587.1 | - | 2 | 266 | |||
BB868845.1 | - | 2 | 344 | |||
BC116976.1 | - | 2 | 270 | 40126295 | ||
BC116978.1 | - | 2 | 354 | 40126297 |
Set Membership: |
![]() |