Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Itgb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016780.2 | NP_058060.2 | 7, 8 | 1119 | ||
GenBank mRNA | AF026509.1 | - | 7, 8 | 1063 | ||
AK156518.1 | - | 3, 4 | 376 | |||
AK157958.1 | - | 7, 8 | 1065 | |||
AK198913.1 | - | 180 | ||||
BC125518.1 | - | 7, 8 | 1119 | 40130033 | ||
BC125520.1 | - | 7, 8 | 1119 | 40130035 |