Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111324.2 | NP_001104794.1 | 3 | 602 | ||
NM_017464.5 | NP_059492.3 | 2 | 383 | |||
GenBank mRNA | AF009366.1 | - | 3 | 538 | ||
AK030985.1 | - | 3 | 536 | |||
AK033729.1 | - | 2 | 384 | |||
AK046357.1 | - | 2 | 346 | |||
AK054179.1 | - | 2 | 379 | |||
AK083374.1 | - | 3 | 530 | |||
BC053713.1 | - | 3 | 535 | 30077589 |