Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mmp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019471.3 | NP_062344.1 | 10 | 1544 | ||
GenBank mRNA | AK020292.1 | - | 10 | 1545 | ||
BC130027.1 | - | 10 | 1536 | 40140858 | ||
Y13185.1 | - | 10 | 1533 |